Please login to your online account to display your discounted pricing

Dimethylformamide - D7, deuteration degree min. 99.5% for NMR spectroscopy MagniSolv™, EMD Millipore™

For NMR Spectroscopy

$120.49 - $712.77

Chemical Identifiers

Chemical Name or Material Dimethylformamide - D7
CAS 4472-41-7
Physical Form Liquid
Assay Percent Range 99.5% min.
Molecular Formula C3D7NO (Hill Formula)
Formula Weight 80.14
Synonym Heptadeuterodimethylformamide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1116560009

 
emd millipore corporation
1116560009
Glass ampoule 10 x 0.75mL Each for $712.77

M1116560001

 
emd millipore corporation
1116560001
Glass ampoule 1mL Each for $120.49
Description & Specifications

Specifications

Chemical Name or Material Dimethylformamide - D7
CAS 4472-41-7
Odor Weakly amine - like
Packaging Glass ampoule
Ignition Point 440°C (824°F)
Vapor Pressure 3.77 hPa at 20°C (68°F)
Physical Form Liquid
Quantity 10 x 0.75mL
Assay Percent Range 99.5% min.
Color Colorless
Molecular Formula C3D7NO (Hill Formula)
Formula Weight 80.14
Synonym Heptadeuterodimethylformamide