missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl trans-1,4-cyclohexanedicarboxylate, 99+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 240790250

 View more versions of this product

Catalog No. AC240790250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Dimethyl trans-1,4-cyclohexanedicarboxylate
Authentic
Glass Bottle
CH3OOCC6H10COOCH3
25g
Solubility in water: practically insoluble. Other solubilities: soluble in most common organic solvents
COC(=O)C1CCC(CC1)C(=O)OC
200.23
200.23
3399-22-2
99% min. (GC)
C10H16O4
MFCD00063917,MFCD00001460,MFCD00216473
dimethyl 1,4-cyclohexanedicarboxylate, dimethyl trans-1,4-cyclohexanedicarboxylate, 1r,4r-dimethyl cyclohexane-1,4-dicarboxylate, cis-dimethyl cyclohexane-1,4-dicarboxylate, dimethyl trans-cyclohexane-1,4-dicarboxylate, 1,4-cyclohexanedicarboxylic acid, dimethyl ester, dimethyl hexahydroterephthalate, trans-dimethyl cyclohexane-1,4-dicarboxylate, 1,4-cyclohexanedicarboxylic acid, dimethyl ester, trans, unii-ys87mds0zv
LNGAGQAGYITKCW-UHFFFAOYSA-N
dimethyl cyclohexane-1,4-dicarboxylate
7198
99+%

Chemical Identifiers

3399-22-2
200.23
LNGAGQAGYITKCW-UHFFFAOYSA-N
7198
COC(=O)C1CCC(CC1)C(=O)OC
C10H16O4
MFCD00063917,MFCD00001460,MFCD00216473
dimethyl 1,4-cyclohexanedicarboxylate, dimethyl trans-1,4-cyclohexanedicarboxylate, 1r,4r-dimethyl cyclohexane-1,4-dicarboxylate, cis-dimethyl cyclohexane-1,4-dicarboxylate, dimethyl trans-cyclohexane-1,4-dicarboxylate, 1,4-cyclohexanedicarboxylic acid, dimethyl ester, dimethyl hexahydroterephthalate, trans-dimethyl cyclohexane-1,4-dicarboxylate, 1,4-cyclohexanedicarboxylic acid, dimethyl ester, trans, unii-ys87mds0zv
dimethyl cyclohexane-1,4-dicarboxylate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only