missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl trans-1,4-cyclohexanedicarboxylate, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC240790010

 View more versions of this product

Catalog No. AC240790010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Dimethyl trans-1,4-cyclohexanedicarboxylate
Glass bottle
67°C to 70°C
3399-22-2
C10H16O4
MFCD00063917
Solubility in water: practically insoluble. Other solubilities: soluble in most common organic solvents
COC(=O)C1CCC(CC1)C(=O)OC
200.23
200.23
99+%
Authentic
White to Yellow
1g
99% min. (GC)
CH3OOCC6H10COOCH3
1,4-cyclohexanedicarboxylic acid, dimethyl ester, 1,4-cyclohexanedicarboxylic acid, dimethyl ester, trans, 1r,4r-dimethyl cyclohexane-1,4-dicarboxylate, cis-dimethyl cyclohexane-1,4-dicarboxylate, dimethyl 1,4-cyclohexanedicarboxylate, dimethyl hexahydroterephthalate, dimethyl trans-1,4-cyclohexanedicarboxylate, dimethyl trans-cyclohexane-1,4-dicarboxylate, trans-dimethyl cyclohexane-1,4-dicarboxylate, unii-ys87mds0zv
LNGAGQAGYITKCW-UHFFFAOYSA-N
dimethyl cyclohexane-1,4-dicarboxylate
7198
Crystalline Powder

Chemical Identifiers

3399-22-2
200.23
LNGAGQAGYITKCW-UHFFFAOYSA-N
7198
COC(=O)C1CCC(CC1)C(=O)OC
C10H16O4
MFCD00063917
1,4-cyclohexanedicarboxylic acid, dimethyl ester, 1,4-cyclohexanedicarboxylic acid, dimethyl ester, trans, 1r,4r-dimethyl cyclohexane-1,4-dicarboxylate, cis-dimethyl cyclohexane-1,4-dicarboxylate, dimethyl 1,4-cyclohexanedicarboxylate, dimethyl hexahydroterephthalate, dimethyl trans-1,4-cyclohexanedicarboxylate, dimethyl trans-cyclohexane-1,4-dicarboxylate, trans-dimethyl cyclohexane-1,4-dicarboxylate, unii-ys87mds0zv
dimethyl cyclohexane-1,4-dicarboxylate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents