Please login to your online account to display your discounted pricing

Dimethylethylhexadecylammonium bromide, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  211870050

 View more versions of this product

Catalog No. AC211870050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Powder
Formula Weight 378.48
Quantity 5g
Color White
EINECS Number 204-672-8
TSCA TSCA
pH 6 to 8 (1% soln.)
RTECS Number BQ5425000
Beilstein 04, III, 425
Merck Index 15, 2028
Name Note 99%
Molecular Formula C20H44BrN
Linear Formula CH3(CH2)15N(CH3)2(C2H5)Br
Infrared Spectrum Authentic
MDL Number MFCD00011774
CAS 124-03-8
Packaging Glass bottle
Chemical Name or Material Dimethylethylhexadecylammonium bromide
Assay Percent Range 99%
Synonym Cetyldimethylethylammonium bromide
Additional Information amine hydrobromide: < 0.5%
Amine < 0.2%
Health Hazard 1 HYGROSCOPIC
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Dimethylethylhexadecylammonium bromide
Name Note 99%
CAS 124-03-8
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C20H44BrN
Linear Formula CH3(CH2)15N(CH3)2(C2H5)Br
Formula Weight 378.48
MDL Number MFCD00011774
Synonym Cetyldimethylethylammonium bromide
Accessories

HYGROSCOPIC

GHS P Statement:
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

SDS & Safety Documents