missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+)-DimethyleL-tartrate, 99%, ACROS Organics™

Manufacturer:  Acros Organics 172490250

 View more versions of this product

Catalog No. AC172490250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(+)-Dimethyl L-tartrate
White
608-68-4
100.0
Glass bottle
≥98.5%
C6H10O6
MFCD00064437
13,124
COC(=O)C(C(C(=O)OC)O)O
178.14
178.14
+ 21.00 (20.00°C c=10,H2O)
25g
98.5
Authentic
+20° to +22° (20°C, 589nm) (c=10, H20)
99%
[-CH(OH)CO2CH3]2
03, 510
PVRATXCXJDHJJN-QWWZWVQMSA-N
dimethyl (2R,3R)-2,3-dihydroxybutanedioate
11851
Crystals

Chemical Identifiers

608-68-4
178.14
PVRATXCXJDHJJN-QWWZWVQMSA-N
C6H10O6
MFCD00064437
11851
SDS Documents