missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethylefumarate, 99%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 222180250

 View more versions of this product

Catalog No. AC222180250


 

Specifications Description & Specifications

Specifications

Dimethylefumarate
100° to 104°C
624-49-7
Liquid
99%
C6H8O4
144.13
02, 741
Clear colorless to faintly yellow
25g
Authentic
≥98.5% (GC)
Glass bottle
CH3O2CCH=CHCO2CH3
MFCD00064438

Chemical Identifiers

99%
CH3O2CCH=CHCO2CH3
MFCD00064438
≥98.5% (GC)
C6H8O4
144.13
Dimethylefumarate

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.