Learn More
Dimethyl chlorophosphate, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 426140250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Dimethyl chlorophosphate | |
1.3400g/mL | |
>110°C | |
Glass bottle | |
(CH3O)2POCl | |
25g | |
dimethyl phosphorochloridate, dimethyl chlorophosphate, phosphorochloridic acid, dimethyl ester, methyl phosphorochloridate, dimethylchlorophosphate, dimethyl chlorophosphonate, chlorodimethoxyphosphine oxide, chlorodimethylphosphate, dimethyl_chlorophosphate, pubchem19321 | |
COP(Cl)(=O)OC | |
144.49 | |
144.49 |
813-77-4 | |
80°C (25.0mmHg) | |
95% min. (Argentometry) | |
C2H6ClO3P | |
MFCD00117904 | |
1.34 | |
NGFFLHMFSINFGB-UHFFFAOYSA-N | |
[chloro(methoxy)phosphoryl]oxymethane | |
101863 | |
96% |
Safety and Handling
GHS H Statement:
Fatal if swallowed.
Causes severe skin burns and eye damage.
Fatal in contact with skin.
Fatal if inhaled.
Contact with water liberates toxic gas.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and
GHS Signal Word: Danger
RUO â Research Use Only