Learn More
Dimethyl carbonate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 116120010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentSpecifications
Dimethyl carbonate | |
2.0°C to 4.0°C | |
90.0°C | |
Authentic | |
Glass bottle | |
1.3672 to 1.3692 | |
MFCD00008420 | |
03, 4 | |
1.069 | |
methyl carbonate, carbonic acid, dimethyl ester, carbonic acid dimethyl ester, methyl carbonate meo 2co, unii-ke9j097spn, dimethylcarbonate, ch3ocooch3, ke9j097spn, dimethyl ester of carbonic acid, dsstox_cid_9192 | |
IEJIGPNLZYLLBP-UHFFFAOYSA-N | |
dimethyl carbonate | |
12021 | |
CHEBI:36596 | |
99% |
616-38-6 | |
1.0690g/mL | |
18°C | |
98.5% min. (GC) | |
C3H6O3 | |
(CH3O)2CO | |
1 kg | |
02,149; 05,234; 12,201 | |
15, 3263 | |
Solubility in water: 139g/L. Other solubilities: soluble in all proportions with organic alcohols,soluble in all proportions with esters and ketones,miscible with acids and alkalies | |
COC(=O)OC | |
90.08 | |
0.625 mPa.s (20°C) | |
90.08 | |
Liquid |
Chemical Identifiers
616-38-6 | |
90.08 | |
IEJIGPNLZYLLBP-UHFFFAOYSA-N | |
12021 | |
dimethyl carbonate |
C3H6O3 | |
MFCD00008420 | |
methyl carbonate, carbonic acid, dimethyl ester, carbonic acid dimethyl ester, methyl carbonate meo 2co, unii-ke9j097spn, dimethylcarbonate, ch3ocooch3, ke9j097spn, dimethyl ester of carbonic acid, dsstox_cid_9192 | |
CHEBI:36596 | |
COC(=O)OC |
Safety and Handling
GHS H Statement:
Highly flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 210-478-4
RUO – Research Use Only