missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl 5-hydroxyisophthalate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC356440100

 View more versions of this product

Catalog No. AC356440100



Description

Description

Specifications

Specifications

Dimethyl 5-hydroxyisophthalate
Glass bottle
162°C to 165°C
99%
98.5% min. (GC)
MFCD00134367
DOSDTCPDBPRFHQ-UHFFFAOYSA-N
dimethyl 5-hydroxybenzene-1,3-dicarboxylate
83065
Powder and/or Lumps
Authentic
White to Beige
10g
13036-02-7
C10H10O5
1,3-benzenedicarboxylic acid, 5-hydroxy-, 1,3-dimethyl ester, 1,3-benzenedicarboxylic acid, 5-hydroxy-, dimethyl ester, 1,3-dimethyl 5-hydroxybenzene-1,3-dicarboxylate, 5-hydroxy-dimethylisophthalate, 5-hydroxyisophthalic acid dimethyl ester, dimethyl 5-hydroxyisophthalate, dimethyl-5-hydroxy-benzene-1,3-dicarbonate, dimethyl5-hydroxyisophthalate, methyl 5-hydroxy-3-methoxycarbonyl benzoate, timtec-bb sbb000491
COC(=O)C1=CC(=CC(O)=C1)C(=O)OC
210.19
210.19
99%

Chemical Identifiers

13036-02-7
210.19
DOSDTCPDBPRFHQ-UHFFFAOYSA-N
83065
COC(=O)C1=CC(=CC(O)=C1)C(=O)OC
C10H10O5
MFCD00134367
1,3-benzenedicarboxylic acid, 5-hydroxy-, 1,3-dimethyl ester, 1,3-benzenedicarboxylic acid, 5-hydroxy-, dimethyl ester, 1,3-dimethyl 5-hydroxybenzene-1,3-dicarboxylate, 5-hydroxy-dimethylisophthalate, 5-hydroxyisophthalic acid dimethyl ester, dimethyl 5-hydroxyisophthalate, dimethyl-5-hydroxy-benzene-1,3-dicarbonate, dimethyl5-hydroxyisophthalate, methyl 5-hydroxy-3-methoxycarbonyl benzoate, timtec-bb sbb000491
dimethyl 5-hydroxybenzene-1,3-dicarboxylate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 235-899-

SDS Documents