Please login to your online account to display your discounted pricing

Dimethyle4,5-imidazoledicarboxylate, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC408370250

 View more versions of this product

Catalog No. AC408370250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Fine Crystalline Powder
Formula Weight 184.15
Quantity 25g
Color White to light brown
Melting Point 197° to 201°C
Chemical Name or Material Dimethyl 4, 5-imidazoledicarboxylate
Molecular Formula C7H8N2O4
MDL Number MFCD00051338
CAS 3304-70-9
Packaging Glass bottle
Assay Percent Range 98%
Health Hazard 1 Warning
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Dimethyl 4, 5-imidazoledicarboxylate
CAS 3304-70-9
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H8N2O4
Formula Weight 184.15
MDL Number MFCD00051338
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents