missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl 4,5-imidazoledicarboxylate, 98%, ACROS Organics™

$108.15 - $108.15

Chemical Identifiers

CAS 3304-70-9
Molecular Formula C7H8N2O4
Molecular Weight (g/mol) 184.151
MDL Number MFCD00051338
InChI Key CUIWFAXEALIQJS-UHFFFAOYSA-N
Synonym 1h-imidazole-4, dimethyl ester, 1h-imidazole-4,5-dicarboxylic acid dimethyl ester, 1h-imidazole-4,5-dicarboxylic acid, 4,5-dimethyl ester, 1h-imidazole-4,5-dicarboxylic acid, dimethyl ester, 4,5-dimethyl 1h-imidazole-4,5-dicarboxylate, dimethyl 4,5-imidazole-dicarboxylate, dimethyl 4,5-imidazoledicarboxylate, dimethyl imidazole-4,5-dicarboxylate, imidazole-4, dimethyl ester, imidazole-4,5-dicarboxylic acid, dimethyl ester
PubChem CID 76810
IUPAC Name dimethyl 1H-imidazole-4,5-dicarboxylate
SMILES COC(=O)C1=C(N=CN1)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC408370050
SDS View Documents
Thermo Fisher Scientific
408370050
5g Glass bottle
Each for $108.15
 
AC408370250
SDS View Documents
Thermo Fisher Scientific
AC408370250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

3304-70-9
184.151
CUIWFAXEALIQJS-UHFFFAOYSA-N
76810
COC(=O)C1=C(N=CN1)C(=O)OC
C7H8N2O4
MFCD00051338
1h-imidazole-4, dimethyl ester, 1h-imidazole-4,5-dicarboxylic acid dimethyl ester, 1h-imidazole-4,5-dicarboxylic acid, 4,5-dimethyl ester, 1h-imidazole-4,5-dicarboxylic acid, dimethyl ester, 4,5-dimethyl 1h-imidazole-4,5-dicarboxylate, dimethyl 4,5-imidazole-dicarboxylate, dimethyl 4,5-imidazoledicarboxylate, dimethyl imidazole-4,5-dicarboxylate, imidazole-4, dimethyl ester, imidazole-4,5-dicarboxylic acid, dimethyl ester
dimethyl 1H-imidazole-4,5-dicarboxylate

Specifications

Authentic
White to Brown
5g
98%
MFCD00051338
CUIWFAXEALIQJS-UHFFFAOYSA-N
dimethyl 1H-imidazole-4,5-dicarboxylate
76810
Fine Crystalline Powder
Dimethyl 4,5-imidazoledicarboxylate
Glass bottle
197.0°C to 201.0°C
3304-70-9
C7H8N2O4
1h-imidazole-4, dimethyl ester, 1h-imidazole-4,5-dicarboxylic acid dimethyl ester, 1h-imidazole-4,5-dicarboxylic acid, 4,5-dimethyl ester, 1h-imidazole-4,5-dicarboxylic acid, dimethyl ester, 4,5-dimethyl 1h-imidazole-4,5-dicarboxylate, dimethyl 4,5-imidazole-dicarboxylate, dimethyl 4,5-imidazoledicarboxylate, dimethyl imidazole-4,5-dicarboxylate, imidazole-4, dimethyl ester, imidazole-4,5-dicarboxylic acid, dimethyl ester
COC(=O)C1=C(N=CN1)C(=O)OC
184.15
184.15
98%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS