Please login to your online account to display your discounted pricing

Dimethyle4,5-imidazoledicarboxylate, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  408370050

 View more versions of this product

Catalog No. AC408370050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Fine Crystalline Powder
Formula Weight 184.15
Quantity 5g
Color White to light brown
Melting Point 197° to 201°C
Health Hazard 1 Warning
Assay Percent Range 98%
CAS 3304-70-9
Chemical Name or Material Dimethyl 4, 5-imidazoledicarboxylate
Molecular Formula C7H8N2O4
MDL Number MFCD00051338
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Dimethyl 4, 5-imidazoledicarboxylate
CAS 3304-70-9
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H8N2O4
Formula Weight 184.15
MDL Number MFCD00051338
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents