Please login to your online account to display your discounted pricing

Dimethyle4-(4-methoxyphenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Dimethyl 4-(4-methoxyphenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
CAS 73257-47-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C18H21NO5
Formula Weight 331.37
MDL Number MFCD00085027
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SEW02190DA

  SDS
fisher scientific company
SEW02190DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SEW02190EA

  SDS
fisher scientific company
SEW02190EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 331.37
Quantity 1g
Assay 97%
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C18H21NO5
MDL Number MFCD00085027
CAS 73257-47-3
Chemical Name or Material Dimethyl 4-(4-methoxyphenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate