Please login to your online account to display your discounted pricing

Dimethyle4-(4-methoxyphenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SEW02190EA

 View more versions of this product

Catalog No. SEW02190EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 331.37
Quantity 10g
Assay 97%
CAS Min % 97.0
CAS 73257-47-3
Chemical Name or Material Dimethyl 4-(4-methoxyphenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C18H21NO5
MDL Number MFCD00085027

Specifications

Chemical Name or Material Dimethyl 4-(4-methoxyphenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
CAS 73257-47-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C18H21NO5
Formula Weight 331.37
MDL Number MFCD00085027