Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyldiphenyltin

Chemical Identifiers

Chemical Name or Material Dimethyldiphenyltin
CAS 1080-43-9
Formula Weight 302.97
MDL Number MFCD00058812
UN Number UN2788
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3961703

  SDS
alfa aesar™
3961703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3961706

  SDS
alfa aesar™
3961706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3961714

  SDS
alfa aesar™
3961714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 302.97
Chemical Name or Material Dimethyldiphenyltin
Physical Form Liquid
MDL Number MFCD00058812
CAS 1080-43-9
Quantity 1g
UN Number UN2788
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.