missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyldioctadecylammonium bromide, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC408261000

 View more versions of this product

Catalog No. AC408261000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Phase transfer catalyst
Specifications

Specifications

Dimethyldioctadecylammonium bromide
Authentic
1.047
151.0°C to 157.0°C
3700-67-2
C38H80BrN
MFCD00041975
1-octadecanaminium, n,n-dimethyl-n-octadecyl-, bromide, 1-octadecanaminium, n,n-dimethyl-n-octadecyl-, bromide 1:1, dimethyldioctadecylammonium bromide, dimethyldioctadecylazanium bromide, dimethyldistearylammonium bromide, distearyl dimethyl ammonium bromide, distearyldimethylammonium bromide, dsab, n,n-dimethyl-n-octadecyloctadecan-1-aminium bromide, unii-z5t47r065a
PSLWZOIUBRXAQW-UHFFFAOYSA-M
dimethyl(dioctadecyl)azanium;bromide
77293
Granular Powder
1.0470g/mL
Glass bottle
White
100g
99.0 to 101% (Argentometry)
(CH3)2N(Br)((CH2)17CH3)2
04, IV, 829
Solubility in water: .. Other solubilities: soluble in acetonitrile,methanol,ethanol,aceton,e,ethyl acetate
CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCCCC.[Br-]
630.96
630.96
99+%

Chemical Identifiers

3700-67-2
630.96
PSLWZOIUBRXAQW-UHFFFAOYSA-M
77293
CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCCCC.[Br-]
C38H80BrN
MFCD00041975
1-octadecanaminium, n,n-dimethyl-n-octadecyl-, bromide, 1-octadecanaminium, n,n-dimethyl-n-octadecyl-, bromide 1:1, dimethyldioctadecylammonium bromide, dimethyldioctadecylazanium bromide, dimethyldistearylammonium bromide, distearyl dimethyl ammonium bromide, distearyldimethylammonium bromide, dsab, n,n-dimethyl-n-octadecyloctadecan-1-aminium bromide, unii-z5t47r065a
dimethyl(dioctadecyl)azanium;bromide
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF

GHS Signal Word: Warning

EINECSNumber : 223-037-6

SDS Documents