Please login to your online account to display your discounted pricing

Dimethyldioctadecylammonium bromide, +99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  408260250

 View more versions of this product

Catalog No. AC408260250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Granular Powder
Formula Weight 630.96
Melting Point >150°C (clear point)
Quantity 25g
Color White
Beilstein 04, IV, 829
Assay 99.0 to 101.0%
Density 1.0470g/mL
Chemical Name or Material Dimethyldioctadecylammonium bromide
Linear Formula (CH3)2N(Br)((CH2)17CH3)2
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Molecular Formula C38H80BrN
MDL Number MFCD00041975
CAS 3700-67-2
Packaging Glass bottle
Health Hazard 1 Warning

Specifications

Chemical Name or Material Dimethyldioctadecylammonium bromide
Assay 99.0 to 101.0%
Assay Percent Range 99+%
Molecular Formula C38H80BrN
Linear Formula (CH3)2N(Br)((CH2)17CH3)2
Formula Weight 630.96
MDL Number MFCD00041975
Accessories

Warning

GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Causes skin irritation.

SDS & Safety Documents