Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyldi-n-octadecylammonium chloride, 96%

Chemical Identifiers

Chemical Name or Material Dimethyldi-n-octadecylammonium chloride
CAS 107-64-2
Molecular Formula C38H80ClN
Linear Formula [CH3(CH2)17]2N(CH3)2Cl
Formula Weight 586.51
MDL Number MFCD00055540
UN Number UN3077
Beilstein 3580288
Synonym Dimethyldistearylammonium chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2378818

 
alfa aesar™
B2378818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2378830

 
alfa aesar™
B2378830
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 586.51
Chemical Name or Material Dimethyldi-n-octadecylammonium chloride
TSCA TSCA
Assay 96%
Molecular Formula C38H80ClN
Linear Formula [CH3(CH2)17]2N(CH3)2Cl
MDL Number MFCD00055540
CAS 107-64-2
UN Number UN3077
Beilstein 3580288
Quantity 50g
Odor Odorless
Synonym Dimethyldistearylammonium chloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number BQ1923000
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H318
Causes serious eye damage.