Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyldi-n-octadecylammonium bromide, 99%

Chemical Identifiers

Chemical Name or Material Dimethyldi-n-octadecylammonium bromide
CAS 3700-67-2
Molecular Formula [CH3(CH2)17]2N(CH3)2Br
Formula Weight 630.97g/mol
MDL Number MFCD00041975
UN Number UN3077
Beilstein 3580323
Synonym Dimethyldistearylammonium bromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1824403

 
alfa aesar™
A18244-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1824406

 
alfa aesar™
A18244-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 152° to 156°C
Quantity 1g
Odor Odorless
Chemical Name or Material Dimethyldi-n-octadecylammonium bromide
CAS 3700-67-2
Assay 99%
Molecular Formula [CH3(CH2)17]2N(CH3)2Br
Formula Weight 630.97g/mol
MDL Number MFCD00041975
UN Number UN3077
Beilstein 3580323
Synonym Dimethyldistearylammonium bromide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECS Number RG0185984
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.