Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethylaminoacetyl chloride hydrochloride, tech. 85%

Chemical Identifiers

Chemical Name or Material Dimethylaminoacetyl chloride hydrochloride
CAS 60853-81-8
Purity Grade Technical Grade
Molecular Formula (CH3)2NCH2COCl·HCl
Formula Weight 158.03
MDL Number MFCD02094025
UN Number UN3261
Beilstein 3980151
Synonym N,N-Dimethylglycinyl chloride hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1847506

  SDS
alfa aesar™
L1847506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 158.03
Chemical Name or Material Dimethylaminoacetyl chloride hydrochloride
MDL Number MFCD02094025
CAS 60853-81-8
UN Number UN3261
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Purity Grade Technical Grade
Assay 85%
Molecular Formula (CH3)2NCH2COCl·HCl
Beilstein 3980151
Quantity 5g
Synonym N,N-Dimethylglycinyl chloride hydrochloride
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Applications
Dimethylaminoacetyl chloride hydrochloride is used as a pharmaceutical intermediate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Store under inert gas. Incompatible with oxidizing agents and bases.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.