Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethylaminoacetonitrile, 98%

Chemical Identifiers

Chemical Name or Material Dimethylaminoacetonitrile
CAS 926-64-7
Molecular Formula C4H8N2
Linear Formula (CH3)2NCH2CN
Formula Weight 84.12
MDL Number MFCD00001890
UN Number UN2378
Beilstein 1735677
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2508522

 
alfa aesar™
B2508522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2508514

 
alfa aesar™
B2508514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 84.12
Chemical Name or Material Dimethylaminoacetonitrile
Assay 98%
Molecular Formula C4H8N2
Linear Formula (CH3)2NCH2CN
Boiling Point 137° to 138°C
MDL Number MFCD00001890
CAS 926-64-7
UN Number UN2378
Beilstein 1735677
Quantity 100g
Flash Point 36°C (96°F)
Odor Recognizable
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H310-H226-H319
Fatal if swallowed.
Fatal in contact with skin.
Flammable liquid and vapour.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P260-P280h-P305+P351+P338-P309-P310-P302+P352
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2-DIMETHYLAMINOACETONITRILE
RTECS Number AL9450000
Safety and Handling

Danger

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310-P302+P352
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

GHS H Statement
H300-H310-H226-H319
Fatal if swallowed.
Fatal in contact with skin.
Flammable liquid and vapour.
Causes serious eye irritation.