Please login to your online account to display your discounted pricing

Dimethylaminoacetaldehyde dimethylacetal 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Dimethylaminoacetaldehyde dimethylacetal
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H15NO2
Linear Formula (CH3)2NCH2CH(OCH3)2
Formula Weight 133.19
MDL Number MFCD00010205
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC390910010

  SDS
thermo fisher scientific
AC390910010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC390910050

  SDS
thermo fisher scientific
AC390910050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 133.19
Quantity 1g
Color Clear colorless to slightly yellow
Density 0.9g/mL
Chemical Name or Material Dimethylaminoacetaldehyde dimethylacetal
Refractive Index 1.4080 to 1.4120 (20°C, 589nm)
Assay Percent Range 97%
Molecular Formula C6H15NO2
Linear Formula (CH3)2NCH2CH(OCH3)2
MDL Number MFCD00010205
CAS 38711-20-5
Health Hazard 1 GHS Signal Word: Danger
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.