Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethylaminoacetaldehyde diethyl acetal, 95%

Chemical Identifiers

Chemical Name or Material Dimethylaminoacetaldehyde diethyl acetal
CAS 3616-56-6
Molecular Formula (CH3)2NCH2CH(OCH2CH3)2
Formula Weight 161.25
MDL Number MFCD00009232
UN Number UN1993
Beilstein 1739149
Synonym 2,2-Diethoxy-N,N-dimethylethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0627206

  SDS
alfa aesar™
L0627206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0627214

  SDS
alfa aesar™
L0627214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 161.25
Chemical Name or Material Dimethylaminoacetaldehyde diethyl acetal
Assay 95%
CAS 3616-56-6
UN Number UN1993
Beilstein 1739149
Synonym 2,2-Diethoxy-N,N-dimethylethylamine
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Molecular Formula (CH3)2NCH2CH(OCH2CH3)2
Boiling Point 164° to 166°C
MDL Number MFCD00009232
Quantity 5g
Flash Point 45°C (113°F)
Odor Amine-like
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.