Learn More
Dimethylamine, 40% w/w aq. soln., Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03145830
Description
Dimethylamine 40% aqueous solution is used as a precursor in the preparation of nitrosodimethylamine. It is also used in paint additives, lubricants, lubricant additives and coating additives. It plays a major role in ion exchange agents and agricultural chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dimethylamine | |
0.890 g/mL | |
Fish-like | |
1.37 | |
250 g | |
605257 | |
dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution | |
ROSDSFDQCJNGOL-UHFFFAOYSA-N | |
N-methylmethanamine | |
674 | |
45.08 | |
Liquid |
124-40-3 | |
15°C (60°F) | |
C2H7N | |
MFCD00008288 | |
UN1160 | |
14,3228 | |
Miscible with water and most organic solvents. | |
CNC | |
45.085 | |
CHEBI:17170 | |
40% w/w aq. soln. |
Chemical Identifiers
124-40-3 | |
45.085 | |
ROSDSFDQCJNGOL-UHFFFAOYSA-N | |
674 | |
N-methylmethanamine |
C2H7N | |
MFCD00008288 | |
dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution | |
CHEBI:17170 | |
CNC |
Safety and Handling
GHS H Statement
H225-H331-H314-H318-H302-H335
Highly flammable liquid and vapor.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302+H332-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIMETHYLAMINE SOLUTION
EINECSNumber : 204-697-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only