missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethylamine, 40% w/w aq. soln.

Manufacturer:  Alfa Aesar™ 31458A5

 View more versions of this product

Catalog No. AA31458A5



Specifications

Specifications

Dimethylamine
15° (60°F)
1.3700
124-40-3
MFCD00008288
605257
dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution
ROSDSFDQCJNGOL-UHFFFAOYSA-N
N-methylmethanamine
674
45.08
0.890g/mL
Fish-like
3kg
C2H7N
UN1160
14,3228
40% w/w aq. soln.
CNC
45.085
CHEBI:17170
Liquid

Chemical Identifiers

124-40-3
45.085
ROSDSFDQCJNGOL-UHFFFAOYSA-N
674
N-methylmethanamine
C2H7N
MFCD00008288
dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution
CHEBI:17170
CNC
Safety and Handling

Safety and Handling

GHS H Statement
H225-H331-H314-H318-H302-H335
Highly flammable liquid and vapor.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIMETHYLAMINE SOLUTION

TSCA : TSCA

SDS Documents