missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethylamine, 2M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 388388000

 View more versions of this product

Catalog No. AC388388000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dimethylamine
2M solution in THF
C2H7N
MFCD00008288
0.8500g/mL
dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution
ROSDSFDQCJNGOL-UHFFFAOYSA-N
N-methylmethanamine
674
45.08
0.85
−17°C
Yellow
124-40-3,109-99-9
NH(CH3)2
About 2M, exact strength on the certificate of analysis
15, 3250
Solubility in water: soluble. Other solubilities: soluble in most organic solvents
CNC
45.085
CHEBI:17170
AcroSeal™ Glass Bottle
800 mL
Solution
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only