Please login to your online account to display your discounted pricing

Dimethylamine, 2M solution in THF, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC388381000

 View more versions of this product

Catalog No. AC388381000

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 45.08
Physical Form Solution
Quantity 100mL
Color Clear slightly yellow
Packaging AcroSeal™ Glass Bottle
Density 0.8500g/mL
Concentration About 2M, exact strength on the certificate of analysis
Merck Index 15, 3250
Linear Formula NH(CH3)2
Health Hazard 1 Danger
CAS 124-40-3
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. May cause respiratory irritation. Suspected of causing cancer. Highly flammable liquid and vapor. May form explosive peroxides.
Molecular Formula C2H7N
Name Note 2M solution in THF
Chemical Name or Material Dimethylamine
CAS 109-99-9
MDL Number MFCD00008288
Health Hazard 3 GHS P Statement Wear eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. Avoid breathing dust/fume/gas/mist/vapors/spray. Keep container tightly closed. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

Specifications

Chemical Name or Material Dimethylamine
Name Note 2M solution in THF
CAS 124-40-3
CAS 109-99-9
Molecular Formula C2H7N
Linear Formula NH(CH3)2
Formula Weight 45.08
MDL Number MFCD00008288
Accessories

Danger

GHS P Statement Wear eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. Avoid breathing dust/fume/gas/mist/vapors/spray. Keep container tightly closed. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

GHS P Statement Causes severe skin burns and eye damage. May cause respiratory irritation. Suspected of causing cancer. Highly flammable liquid and vapor. May form explosive peroxides.

SDS & Safety Documents