missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dimethylamine, 26 wt.% solution in water, Thermo Scientific Chemicals
$75.22 - $75.22
Specifications
Chemical Name or Material | Dimethylamine |
---|---|
CAS | 124-40-3,7732-18-5 |
Melting Point | -37.0°C |
Color | Colorless |
Density | 0.8900g/mL |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC408000010
|
Thermo Scientific Chemicals
408000010 |
1 kg |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dimethylamine | |
-37.0°C | |
0.8900g/mL | |
−18°C | |
(CH3)2NH | |
MFCD00008288 | |
07,119 | |
0.89 | |
Solubility in water: >500g/L (20°C). Other solubilities: soluble in alcohol and ether | |
CNC | |
45.07 | |
CHEBI:17170 | |
25 to 27% |
124-40-3,7732-18-5 | |
Colorless | |
54.0°C | |
C2H7N | |
1.37 | |
04,39 | |
15,325 | |
dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution | |
ROSDSFDQCJNGOL-UHFFFAOYSA-N | |
N-methylmethanamine | |
674 | |
45.07 | |
Liquid |
RUO – Research Use Only