Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl tetrachloroterephthalate, 97%

$244.09 - $244.09

Chemical Identifiers

Chemical Name or Material Dimethyl tetrachloroterephthalate
CAS 1861-32-1
Molecular Formula C10H6Cl4O4
Formula Weight 331.97
MDL Number MFCD00014902
Beilstein 1888840
Merck Index 14,2839
Synonym Dacthal; DCPA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0880014

  SDS
alfa aesar™
L0880014
25g Each for $244.09
Description & Specifications

Specifications

Formula Weight 331.97
MDL Number MFCD00014902
CAS 1861-32-1
Chemical Name or Material Dimethyl tetrachloroterephthalate
Assay 97%
Molecular Formula C10H6Cl4O4
Melting Point 156° to 159°C
Beilstein 1888840
Synonym Dacthal; DCPA
RTECS Number WZ1500000
Quantity 25g
Merck Index 14,2839