Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl terephthalate-2,3,5,6-d4, 98 atom % D

$92.53 - $92.53

Chemical Identifiers

Chemical Name or Material Dimethyl terephthalate-2,3,5,6-d4
CAS 74079-01-9
Molecular Formula C10D4H6O4
Formula Weight 198.21
MDL Number MFCD00182544
Synonym 1,4-Benzene-2,3,5,6-d4-dicarboxylic acid, dimethyl ester; Dimethyl terephthalate-d4(ring-d4)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH51104MD

  SDS
alfa aesar™
H51104MD
250mg Each for $92.53

AAH5110403

  SDS
alfa aesar™
H5110403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Dimethyl terephthalate-2,3,5,6-d4
CAS 74079-01-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 95%
Assay Percent Notes 98 atom % D
Molecular Formula C10D4H6O4
Formula Weight 198.21
MDL Number MFCD00182544
Quantity 250mg
Synonym 1,4-Benzene-2,3,5,6-d4-dicarboxylic acid, dimethyl ester; Dimethyl terephthalate-d4(ring-d4)
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.