missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl sulfoxide, Spectrophotometric Grade, >99%, Alfa Aesar™

Manufacturer:  Alfa Aesar 32434K7

 View more versions of this product

Catalog No. AA32434K7



Specifications

Specifications

Dimethyl sulfoxide
Sprectrophotometric
>99.9%
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
methylsulfinylmethane
679
78.13
Mild
Colorless
67-68-5
Colorless liquid
C2H6OS
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
CS(=O)C
78.129
CHEBI:28262
Liquid
2.5 hPa (2mm Hg) at 20°C
4L

Chemical Identifiers

67-68-5
78.129
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
679
methylsulfinylmethane
C2H6OS
MFCD00002089
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
CHEBI:28262
CS(=O)C
Safety and Handling

Safety and Handling

H227: Combustible liquid.

EINECSNumber : 200-664-3

SDS Documents