missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl sulfoxide, HPLC grade, >99.9%, Alfa Aesar™

Manufacturer:  Alfa Aesar 22914K2

 View more versions of this product

Catalog No. AA22914K2



Specifications

Specifications

Dimethyl sulfoxide
HPLC Grade
>99.9%
MFCD00002089
CS(=O)C
78.129
CHEBI:28262
Liquid
270°C (518°F)
87°C (188.6°F)
Colorless
1L
67-68-5
Colorless liquid
C2H6OS
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
methylsulfinylmethane
679
78.13
1.101g/cm3
Mild
189°C
18.4°C

Chemical Identifiers

67-68-5
78.129
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
CHEBI:28262
CS(=O)C
C2H6OS
MFCD00002089
679
methylsulfinylmethane
Safety and Handling

Safety and Handling

H227: Combustible liquid.

EINECSNumber : 200-664-3

SDS Documents