Please login to your online account to display your discounted pricing

Dimethyl Sulfoxide, For Spectroscopy Uvasol™, EMD Millipore

For spectroscopy Uvasol™

$264.09 - $992.14

Chemical Identifiers

Chemical Name or Material Dimethyl Sulfoxide
CAS 67-68-5
Molecular Formula C2H6OS
Formula Weight 78.13
Synonym Methanesulfinylmethane, Methyl sulfoxide, Dimethyl(oxido)sulfur, DMSO
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1029500500

 
emd millipore corporation
1029500500
Glass Bottle 500mL Each for $264.09

M1029502500

 
emd millipore corporation
1029502500
Glass Bottle 2.5L Each for $992.14
Description & Specifications

Specifications

Boiling Point 189°C
Melting Point 18.5°C
Formula Weight 78.13
Quantity 500mL
Chemical Name or Material Dimethyl Sulfoxide
Physical Form Liquid
Color Colorless
Molecular Formula C2H6OS
Density 1.10g/cm3 at 20°C
Ignition Point 300°to302°C (572°to576°F)
Odor Charactristic
Packaging Glass Bottle
Refractive Index 1.48
Storage Information 15° to 25°C (59° to 77°F); Tightly closed. Dry
Vapor Pressure 0.6 hPa (20°C)
Flash Point 87°C (189°F)
CAS 67-68-5
Synonym Methanesulfinylmethane, Methyl sulfoxide, Dimethyl(oxido)sulfur, DMSO
Solubility Information Soluble in water: 1000g/L at 20°C
Description The Uvasol™ solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The Uvasol™ solvents offer best UV transmittance.