Please login to your online account to display your discounted pricing

Dimethyl Sulfoxide, Fisher BioReagents™


Quantity 4L
Packaging Glass Bottle

Manufacturer: fisher scientific company  FLBP2314

 View more versions of this product

Catalog No. BP231-4

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 189°C
Formula Weight 78.13
Melting Point 18.4°C
Quantity 4L
Merck Index 15, 3285
Vapor Pressure 0.55mbar at 20°C
Viscosity 1.98 mPa/s at 25°C
Decomposition Information >190°C
Physical Form Liquid
Color Colorless
Assay ≥99.7%
Density 1.100g/cm³
Water 0.2% max.
Assay Percent Range ≥99.7 %
Absorbance ≤0.30 Absorbance Units at 280nm
Identification Pass Test
Additional Information Vapor Pressure: 0.55mbar at 20°C
Packaging Glass Bottle
Residue after Evaporation 0.001% max.
CAS 67-68-5
ChemAlert Storage Symbol Gray
Synonym DMSO
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
Combustible liquid. DMSO readily penetrates skin and may carry other dissolved chemicals into the body. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:2
Instability:0
Recommended Storage RT
Chemical Name or Material Dimethyl Sulfoxide
Molecular Formula C2H6OS
Linear Formula (CH3)2SO

Specifications

Chemical Name or Material Dimethyl Sulfoxide
CAS 67-68-5
Physical Form Liquid
Assay ≥99.7%
Assay Percent Range ≥99.7 %
Molecular Formula C2H6OS
Linear Formula (CH3)2SO
Formula Weight 78.13
Synonym DMSO
Accessories
 

Emergency Overview
Combustible liquid. DMSO readily penetrates skin and may carry other dissolved chemicals into the body. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:2
Instability:0

WARNING!

RT