missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl sulfoxide - D6, w/TMS (0.1vol.%), deuteratn. deg. min.99.9% for NMR MagniSolv™, MilliporeSigma™

For NMR Spectroscopy

$199.00 - $600.95

Chemical Identifiers

CAS 2206-27-1
Molecular Formula C2H6OS
Molecular Weight (g/mol) 84.166
InChI Key IAZDPXIOMUYVGZ-WFGJKAKNSA-N
Synonym deuterated dmso
PubChem CID 75151
IUPAC Name trideuterio(trideuteriomethylsulfinyl)methane
SMILES CS(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
M1035870025 View Documents Emd Millipore Corporation
1035870025
25mL Glass bottle Each for $199.00
 
M1035870100 View Documents Emd Millipore Corporation
1035870100
100mL Glass bottle Each for $600.95
 
Specifications

Chemical Identifiers

2206-27-1
84.166
deuterated dmso
trideuterio(trideuteriomethylsulfinyl)methane
C2H6OS
IAZDPXIOMUYVGZ-WFGJKAKNSA-N
75151
CS(=O)C

Specifications

270°C (518°F)
Glass bottle
Colorless
2206-27-1
C2H6OS
IAZDPXIOMUYVGZ-WFGJKAKNSA-N
trideuterio(trideuteriomethylsulfinyl)methane
75151
Liquid
Characteristic
2.5 hPa at 20°C (68°F)
25mL
99.9% min.
deuterated dmso
CS(=O)C
84.166
84.17
Dimethyl sulfoxide - D6, w/TMS (0.1vol.%)