Please login to your online account to display your discounted pricing

Dimethyl sulfoxide - D6,w/TMS(0.03 vol.%),deuteratn. deg.min. 99.96% for NMR MagniSolv™, EMD Millipore™

For NMR Spectroscopy

$209.79 - $680.67

Chemical Identifiers

Chemical Name or Material Dimethyl sulfoxide - D6
Name Note w/TMS (0.03vol.%)
CAS 2206-27-1
Physical Form Liquid
Assay Percent Range 99.96% min.
Molecular Formula C2D6OS (Hill Formula)
Formula Weight 84.17
Synonym DMSO deuterated
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1035920025

 
emd millipore corporation
1035920025
Glass bottle 25mL Each for $680.67

M1035920005

 
emd millipore corporation
1035920005
Glass ampoule 5mL Each for $209.79
Description & Specifications

Specifications

Chemical Name or Material Dimethyl sulfoxide - D6
Name Note w/TMS (0.03vol.%)
CAS 2206-27-1
Odor Characteristic
Packaging Glass bottle
Ignition Point 270°C (518°F)
Vapor Pressure 2.5 hPa at 20°C (68°F)
Physical Form Liquid
Quantity 25mL
Assay Percent Range 99.96% min.
Color Colorless
Molecular Formula C2D6OS (Hill Formula)
Formula Weight 84.17
Synonym DMSO deuterated