missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl sulfoxide - D6, deuteration deg. min. 99.96% for NMR spectroscopy, MagniSolv™, EMD Millipore™

For NMR Spectroscopy

$197.27 - $652.65

Chemical Identifiers

CAS 2206-27-1
Molecular Formula C2H6OS
Molecular Weight (g/mol) 84.166
InChI Key IAZDPXIOMUYVGZ-WFGJKAKNSA-N
Synonym dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide
PubChem CID 75151
IUPAC Name trideuterio(trideuteriomethylsulfinyl)methane
SMILES CS(=O)C
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
M1035620005 View Documents Millipore Sigma
1035620005
10 x 0.5mL Glass ampoule Each for $197.27
M1035620009 View Documents Millipore Sigma
1035620009
10 x 0.75mL Glass ampoule Each for $297.05
M1035620010 View Documents Millipore Sigma
1035620010
10mL Glass ampoule Each for $327.20
M1035620025 View Documents Millipore Sigma
1035620025
25mL Glass bottle Each for $652.65
Specifications

Chemical Identifiers

2206-27-1
84.166
dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide
trideuterio(trideuteriomethylsulfinyl)methane
C2H6OS
IAZDPXIOMUYVGZ-WFGJKAKNSA-N
75151
CS(=O)C

Specifications

270°C (518°F)
Glass ampoule
Colorless
2206-27-1
C2H6OS
IAZDPXIOMUYVGZ-WFGJKAKNSA-N
trideuterio(trideuteriomethylsulfinyl)methane
75151
Liquid
Characteristic
2.5 hPa at 20°C (68°F)
10 x 0.5mL
99.96% min.
dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide
CS(=O)C
84.166
84.17
Dimethyl sulfoxide - D6