Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl sulfoxide, Bioreagent


Quantity 50mL

Manufacturer: alfa aesar™  J66650AD

 View more versions of this product

Catalog No. AAJ66650AD

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 78.13
MDL Number MFCD00002089
Physical Form Liquid
Chemical Name or Material Dimethyl sulfoxide
RTECS Number PV6210000
Health Hazard 1 Warning
Color Clear, colorless
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 506008
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Quantity 50mL
Boiling Point 189°C
TSCA TSCA
Synonym DMSO
Melting Point 18.4°C
Odor Mild
Density 1.101g/mL
Sensitivity Hygroscopic
Name Note Bioreagent
Refractive Index 1.4790
CAS 67-68-5

Specifications

Chemical Name or Material Dimethyl sulfoxide
CAS 67-68-5
Name Note Bioreagent
Formula Weight 78.13
MDL Number MFCD00002089
Beilstein 506008
Synonym DMSO

Applications
DMSO is a polar aprotic solvent used in chemical reactions, in polymerase chain reactions and as a cryoprotectant vitrification agent for the preservation of cells, tissues and organs. DMSO is used in cell freezing media to protect cells from ice crystal induced mechanical injury. It is used for frozen storage of primary, sub-cultured, and recombinant heteroploid and hybridoma cell lines, embryonic stem cells and hematopoietic stem cells. DMSO is frequently used in the combinations with BSA or fetal bovine serum.

Solubility
Soluble in water, methanol, acetone, ether, benzene, chloroform.

Notes
Submicron filtered. Store at Room Temperature (15-30°C), desiccate. Hygroscopic.

Accessories

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.

SDS & Safety Documents