missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl sulfoxide, anhydrous, >99.8%, Alfa Aesar™

Manufacturer:  Alfa Aesar 43998M1

 View more versions of this product

Catalog No. AA43998M1



Specifications

Specifications

Dimethyl sulfoxide
>99.8%
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
methylsulfinylmethane
679
78.13
270°C (518°F)
2mm Hg at 20°C
189°C
18.4°C
67-68-5
C2H6OS
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
CS(=O)C
78.129
CHEBI:28262
Liquid
Mild
87°C (188.6°F)
Colorless
4 x 1L

Chemical Identifiers

67-68-5
78.129
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
679
methylsulfinylmethane
C2H6OS
MFCD00002089
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
CHEBI:28262
CS(=O)C
Safety and Handling

Safety and Handling

H227: Combustible liquid.

EINECSNumber : 200-664-3

Documents