missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl sulfide, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 022949K2

 View more versions of this product

Catalog No. AA22949K2



Description

Description

Dimethyl sulfide is used as a displaceable ligand in chloro(dimethyl sulfide)gold(I) and other coordination compounds, as a reducing agent in ozonolysis reactions as well as a food flavoring component. Further, it is utilized for the preparation of dimethyl sulfoxide, which is a common and important industrial solvent. It finds application in the petroleum industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dimethyl sulfide
-83°C
38°C
Unpleasant
1.4351
1 L
14,6123
Miscible with alcohols,ethers,esters,ketones,aliphatic and aromatic hydrocarbons. Slightly miscible with
CSC
62.13
CHEBI:17437
≥99%
75-18-3
0.845 g/mL
-48°C (-54°F)
C2H6S
MFCD00008562
UN1164
dimethyl sulfide, methyl sulfide, methane, thiobis, dimethylsulfide, dimethylsulphide, methyl thioether, 2-thiapropane, dimethyl sulphide, dimethyl thioether, methylthiomethane
QMMFVYPAHWMCMS-UHFFFAOYSA-N
methylsulfanylmethane
1068
62.14
Liquid

Chemical Identifiers

75-18-3
62.13
QMMFVYPAHWMCMS-UHFFFAOYSA-N
1068
CSC
C2H6S
MFCD00008562
dimethyl sulfide, methyl sulfide, methane, thiobis, dimethylsulfide, dimethylsulphide, methyl thioether, 2-thiapropane, dimethyl sulphide, dimethyl thioether, methylthiomethane
CHEBI:17437
Safety and Handling

Safety and Handling

GHS H Statement
H225-H318-H302-H335-H315
Highly flammable liquid and vapor.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c

H225-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIMETHYL SULFIDE

EINECSNumber : 200-846-2

RTECSNumber : PV5075000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only