Please login to your online account to display your discounted pricing

Dimethyl suberate, 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  165041000

 View more versions of this product

Catalog No. AC165041000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 202.25
Physical Form Liquid
Quantity 100g
Color Clear colorless to slightly yellow
Beilstein 02, 693
TSCA TSCA
EINECS Number 217-059-5
Density 1.01g/mL
Refractive Index 1.4325 to 1.4345 (20°C, 589nm)
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym Suberic acid dimethylester
Chemical Name or Material Dimethyl suberate
MDL Number MFCD00008471
Assay Percent Range 99%
Name Note 99%
Linear Formula CH3O2C(CH2)6CO2CH3
CAS 1732-09-8
Molecular Formula C10H18O4

Specifications

Chemical Name or Material Dimethyl suberate
Name Note 99%
CAS 1732-09-8
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H18O4
Linear Formula CH3O2C(CH2)6CO2CH3
Formula Weight 202.25
MDL Number MFCD00008471
Synonym Suberic acid dimethylester