Please login to your online account to display your discounted pricing

Dimethyl suberate, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  165040250

 View more versions of this product

Catalog No. AC165040250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥98.5%
Formula Weight 202.25
Quantity 25g
Color Clear colorless to slightly yellow
Beilstein 02, 693
EINECS Number 217-059-5
TSCA TSCA
Density 1.01g/mL
Chemical Name or Material Dimethyl suberate
Packaging Glass bottle
Refractive Index 1.4325 to 1.4345 (20°C, 589nm)
Name Note 99%
Assay Percent Range 99%
Molecular Formula C10H18O4
Linear Formula CH3O2C(CH2)6CO2CH3
MDL Number MFCD00008471
CAS 1732-09-8
Synonym Suberic acid dimethylester
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Dimethyl suberate
Name Note 99%
CAS 1732-09-8
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H18O4
Linear Formula CH3O2C(CH2)6CO2CH3
Formula Weight 202.25
MDL Number MFCD00008471
Synonym Suberic acid dimethylester