Please login to your online account to display your discounted pricing

Dimethyl suberate, 99%, ACROS Organics™

$213.98 - $644.63

Chemical Identifiers

Chemical Name or Material Dimethyl suberate
Name Note 99%
CAS 1732-09-8
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H18O4
Linear Formula CH3O2C(CH2)6CO2CH3
Formula Weight 202.25
MDL Number MFCD00008471
Synonym Suberic acid dimethylester
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC165041000

  SDS
thermo fisher scientific
165041000
Glass bottle 100g Each for $213.98

AC165040250

  SDS
fisher scientific acros
165040250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC165045000

  SDS
thermo fisher scientific
165045000
Glass bottle 500g Each for $644.63
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥98.5%
Formula Weight 202.25
Quantity 100g
Color Clear colorless to slightly yellow
Beilstein 02, 693
EINECS Number 217-059-5
TSCA TSCA
Density 1.01g/mL
Chemical Name or Material Dimethyl suberate
Packaging Glass bottle
Refractive Index 1.4325 to 1.4345 (20°C, 589nm)
Name Note 99%
Assay Percent Range 99%
Molecular Formula C10H18O4
Linear Formula CH3O2C(CH2)6CO2CH3
MDL Number MFCD00008471
CAS 1732-09-8
Synonym Suberic acid dimethylester
Infrared Spectrum Authentic