missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl selenide

Manufacturer:  Alfa Aesar™ 6811603

 View more versions of this product

Catalog No. AA6811603



Specifications

Specifications

56°C to 58°C
1g
1.41g/mL
Air & Light Sensitive
C2H6Se
MFCD00014848
1696848
RVIXKDRPFPUUOO-UHFFFAOYSA-N
methylselanylmethane
11648
109.03
Colorless
Dimethyl selenide
Unpleasant
593-79-3
(CH3)2Se
UN3282
4-01-00-01288 beilstein handbook reference, dimethyl selenide gc, methane selenobis-9ci, methylselanyl methane, dimethyl selenide 1g, selenobismethane 9ci, methyl selenide 8ci, selenide dimethyl
C[Se]C
109.041
CHEBI:4610
Liquid

Chemical Identifiers

593-79-3
109.041
RVIXKDRPFPUUOO-UHFFFAOYSA-N
11648
methylselanylmethane
C2H6Se
MFCD00014848
4-01-00-01288 beilstein handbook reference, dimethyl selenide gc, methane selenobis-9ci, methylselanyl methane, dimethyl selenide 1g, selenobismethane 9ci, methyl selenide 8ci, selenide dimethyl
CHEBI:4610
C[Se]C
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P261-P301+P310-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOMETALLIC COMPOUND, TOXIC, LIQUID, N.O.S.

RTECSNumber : VS6880000

Recommended Storage : 2°C to 8°C

SDS Documents