Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl (phthalimidomethyl)phosphonate, 97%

$59.38 - $670.51

Chemical Identifiers

Chemical Name or Material Dimethyl (phthalimidomethyl)phosphonate
CAS 28447-26-9
Molecular Formula C11H12NO5P
Formula Weight 269.19
MDL Number MFCD00015464
Beilstein 1476370
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0284004

  SDS
alfa aesar™
L0284004
2g Each for $59.38

AAL0284009

  SDS
alfa aesar™
L0284009
10g Each for $179.31

AAL0284018

  SDS
alfa aesar™
L0284018
50g Each for $670.51
Description & Specifications

Specifications

Formula Weight 269.19
CAS 28447-26-9
Chemical Name or Material Dimethyl (phthalimidomethyl)phosphonate
Molecular Formula C11H12NO5P
Assay 97%
Beilstein 1476370
Quantity 2g
Melting Point 115° to 120°C
MDL Number MFCD00015464