missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dimethyl phthalate, 99%

Manufacturer:  Alfa Aesar A1425336

 View more versions of this product

Catalog No. AAA1425336


 

Description

Description

Applications
Dimethyl phthalate is used as a plasticizer, solvent, rubber coating agent, molding powder, finishing agent and intermediate. Further, it is used in solid rocket propellants, safety glass, lacquer coatings, adhesives and sealants, toner and colorant products.

Solubility
Miscible with alcohol, ethanol, ether, benzene, chloroform, esters, ketones, organic solvents and chlorinated hydrocarbons. Slightly miscible with carbon tetrachloride. Immiscible with water, petroleum ether and paraffin hydrocarbons.

Notes
Incompatible with oxidizing agents and acids.
Specifications

Specifications

Dimethyl Phthalate
500g
99%
C10H10O4
1911460
NIQCNGHVCWTJSM-UHFFFAOYSA-N
dimethyl benzene-1,2-dicarboxylate
8554
194.19
1.190
131-11-3
99%
MFCD00008425
14,3255
COC(=O)C1=CC=CC=C1C(=O)OC
194.186
CHEBI:4609
Liquid

Chemical Identifiers

131-11-3
194.186
NIQCNGHVCWTJSM-UHFFFAOYSA-N
CHEBI:4609
C10H10O4
MFCD00008425
8554
Safety and Handling

Safety and Handling

RTECSNumber : TI1575000

TSCA : TSCA

Documents