missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dimethyl Phthalate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 116640250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Dimethyl phthalate | |
1.1900g/mL | |
150°C | |
98.5% min. (GC) | |
C10H10O4 | |
MFCD00008425 | |
09, 797 | |
15, 3279 | |
Solubility in water: negligible. Other solubilities: miscible with organic solvents,insoluble in paraffinic hydrocarbons,slightly soluble in mineral oil | |
COC(=O)C1=CC=CC=C1C(=O)OC | |
194.19 | |
17 mPa.s (20°C) | |
194.19 |
131-11-3 | |
282°C | |
Authentic | |
Metal drum | |
1.5140 to 1.516 | |
25L | |
1.19 | |
dimethyl phthalate, dimethylphthalate, solvanom, solvarone, avolin, fermine, phthalic acid dimethyl ester, mipax, palatinol m, unimoll dm | |
NIQCNGHVCWTJSM-UHFFFAOYSA-N | |
dimethyl benzene-1,2-dicarboxylate | |
8554 | |
CHEBI:4609 | |
99% |
Safety and Handling
EINECSNumber : 205-011-6
RUO â Research Use Only