Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl phosphite, 98%

Chemical Identifiers

Chemical Name or Material Dimethyl phosphite
CAS 868-85-9
Molecular Formula C2H7O3P
Formula Weight 110.05
MDL Number MFCD00044633
UN Number UN3278
Beilstein 1697490
Synonym Phosphorous acid dimethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL02670AE

 
alfa aesar™
L02670AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL02670AP

 
alfa aesar™
L02670AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 110.05
CAS 868-85-9
Chemical Name or Material Dimethyl phosphite
Assay ≥98%
Molecular Formula C2H7O3P
MDL Number MFCD00044633
UN Number UN3278
Synonym Phosphorous acid dimethyl ester
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H311-H351-H227-H315-H319-H335
Toxic in contact with skin.
Suspected of causing cancer.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number SZ7710000
Flash Point 70°C (158°F)
Odor Unpleasant
Boiling Point 170° to 171°C
Quantity 100mL
TSCA TSCA
Beilstein 1697490
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.

Applications
Dimethyl phosphite is used as an anticombustioner and water quality stabilizer. It is lubricating oil additives, adhesives.

Solubility
Soluble in water.

Notes
Incompatible with water, strong oxidizing agents, acid chlorides, strong bases. Moisture Sensitive.

Safety and Handling

Danger

GHS P Statement
P210-P261-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H311-H351-H227-H315-H319-H335
Toxic in contact with skin.
Suspected of causing cancer.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.