Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl perfluorosuberate, 96%

$78.04 - $78.04

Chemical Identifiers

Chemical Name or Material Dimethyl perfluorosuberate
CAS 2062-20-6
Molecular Formula C10H6F12O4
Linear Formula CH3O2C(CF2)6CO2CH3
Formula Weight 418.14
MDL Number MFCD00044088
Beilstein 1917354
Synonym Dimethyl dodecafluorooctanedioate; Perfluorosuberic acid dimethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1675406

  SDS
alfa aesar™
L1675406
5g Each for $78.04
Description & Specifications

Specifications

Formula Weight 418.14
Chemical Name or Material Dimethyl perfluorosuberate
Assay 96%
Molecular Formula C10H6F12O4
Melting Point 26°C
Linear Formula CH3O2C(CF2)6CO2CH3
MDL Number MFCD00044088
Synonym Dimethyl dodecafluorooctanedioate; Perfluorosuberic acid dimethyl ester
Boiling Point 152°C (60mmHg)
CAS 2062-20-6
Beilstein 1917354
Quantity 5g