Please login to your online account to display your discounted pricing

Alfa Aesar™ Dimethyl perfluoro-1,10-decanedicarboxylate, 95%

$100.81 - $100.81

Chemical Identifiers

Chemical Name or Material Dimethyl perfluoro-1,10-decanedicarboxylate
CAS 84750-88-9
Molecular Formula C14H6F20O4
Linear Formula CH3O2C(CF2)10CO2CH3
Formula Weight 618.17
MDL Number MFCD00236606
Synonym Eicosafluorododecanedioic acid dimethyl ester; Perfluoro-1,10-decanedicarboxylic acid dimethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1675306

  SDS
alfa aesar™
L1675306
5g Each for $100.81
Description & Specifications

Specifications

Formula Weight 618.17
CAS 84750-88-9
Chemical Name or Material Dimethyl perfluoro-1,10-decanedicarboxylate
Assay 95%
MDL Number MFCD00236606
Synonym Eicosafluorododecanedioic acid dimethyl ester; Perfluoro-1,10-decanedicarboxylic acid dimethyl ester
Melting Point 33° to 40°C
Linear Formula CH3O2C(CF2)10CO2CH3
Quantity 5g
Molecular Formula C14H6F20O4